missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSNAXIP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17730-1-AP-20UL
This item is not returnable.
View return policy
Description
Possible role in spermatogenesis.Specifications
| TSNAXIP1 | |
| Polyclonal | |
| Unconjugated | |
| TSNAXIP1 | |
| Trax interacting protein 1, TSNAXIP1, TXI1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55815, 72236 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q2TAA8, Q99P25 | |
| TSNAXIP1 | |
| TSNAXIP1 Fusion Protein Ag12130 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction