missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSLPR/CRLF2 Antibody (59N5G4) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-27331JF635
This item is not returnable.
View return policy
Description
TSLPR/CRLF2 Monoclonal antibody specifically detects TSLPR/CRLF2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| TSLPR/CRLF2 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CRL2cytokine receptor CRL2 precusor, CRLF2Y, Cytokine receptor-like 2, cytokine receptor-like factor 2, ILXR, IL-XR, P2RY8/CRLF2 fusion, Thymic stromal lymphopoietin protein receptor, thymic stromal lymphopoietin receptor, thymic stromal-derived lymphopoietin receptor, TSLP receptor, TSLPR | |
| Amino acids 119-231 of human TSLPR protein were used as the immunogen for the antibody. | |
| 0.1 mL | |
| Immunology | |
| 64109 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| 59N5G4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction