missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSLP Receptor Polyclonal Antibody

Description
Despite its predicted molecular weight, TSLP-R often migrates at a higher molecular weight in SDS-PAGE. A suggested positive control is human liver tissue lysate. PA5-20378 can be used with blocking peptide PEP-0495.
Thymic stromal lymphopoietin (TSLP) has recently been identified as an important factor capable of driving dendritic cell maturation and activation. It is involved in the positive selection of regulatory T cells, maintenance of peripheral CD4+ T cell homeostasis and the induction of CD4+ T cell-mediated allergic reaction. TSLP is also capable of supporting the growth of fetal liver and adult B cell progenitors and their differentiation to the IgM-positive stage of B cell development. Its receptor TSLP-R will bind TSLP in a low-affinity fashion in transfected cells; co-transfection with IL-7R-alpha cDNA results in high-affinity binding and a functional heteromeric complex. This heteromeric receptor requires stat5 for TSLP-mediated signal transduction and is inhibited by SOCS-1.
Specifications
Specifications
| Antigen | TSLP Receptor |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | CRLF2 |
| Gene Accession No. | Q9HC73 |
| Gene Alias | CRL2; Crlf2; CRLF2Y; Crlm2; CRLM-2; cytokine receptor CRL2 precusor; Cytokine receptor-like 2; Cytokine receptor-like factor 2; cytokine receptor-like molecule 2; delta 1; ILXR; IL-XR; Ly114; lymphocyte antigen 114; P2RY8/CRLF2 fusion; thymic stromal lymphopoietin protein receptor; thymic stromal-derived lymphopoietin receptor; thymic stromal-derived lymphopoietin, receptor; Tpte2; transmembrane phosphoinositide 3-phosphatase and tensin homolog 2; TSLP receptor; Tslpr; type I cytokine receptor delta 1; WI2-87105E3.1 |
| Gene Symbols | CRLF2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?