missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSLP Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13778-1-AP-150UL
This item is not returnable.
View return policy
Description
TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintece of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.Specifications
| TSLP | |
| Polyclonal | |
| Unconjugated | |
| TSLP | |
| thymic stromal lymphopoietin, TSLP | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 85480 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q969D9 | |
| TSLP | |
| TSLP Fusion Protein Ag4771 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction