missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSLP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA585586
This item is not returnable.
View return policy
Description
Keep as concentrated solution. Positive Control: PC-3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TSLP is a 159 amino acid four helix-bundle hemopoietic cytokine that signals through a heterodimeric receptor complex consisting of the human TSLP receptor and the IL-7R alpha-chain. It impacts myeloid cells and induces the release of T cell-attracting chemokines from monocytes, enhances the maturation of CD11c (+) dendritic cells, naive CD4 (+), and CD8 (+) T cells into proallergic effectors, and primes dendritic cells to produce large amounts of IL-12 after CD40 ligand stimulation. TSLP influences the regulation of the positive selection of regulatory T cells and maintenance of peripheral CD4 (+) T cell homeostasis. It induces allergic inflammation by directly activating mast cells and may be involved in the pathophysiology of inflammatory arthritis. It is expressed in heart, liver and prostate.
Specifications
| TSLP | |
| Polyclonal | |
| Unconjugated | |
| TSLP | |
| Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 85480 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q969D9 | |
| TSLP | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human TSLP. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction