missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSLP Monoclonal Antibody (K49024_1G12), Invitrogen™
Mouse Monoclonal Antibody
£380.00 - £570.00
Specifications
| Antigen | TSLP |
|---|---|
| Clone | K49024_1G12 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30211491
|
Invitrogen™
MA556148 |
200 μL |
£570.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30203631
|
Invitrogen™
MA555768 |
50 μL |
£380.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sequence of this protein is as follows: YDFTNCDFEK IKAAYLSTIS KDLITYMSGT KSTEFNNTVS CSNRPHCLTE IQSLTFNPTA GCASLAKEMF AMKTKAALAI WCPGYSETQI NATQAMKKRR KRKVTTNKCL EQVSQLQGLW RRFNRPLLKQ Q
TSLP Monoclonal antibody specifically detects TSLP in Human samples. It is validated for Western BlotSpecifications
| TSLP | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Thymic stroma-derived lymphopoietin; thymic stromal lymphopoietin; TSLP | |
| Recombinant human TSLP (immunogen range is 29-159). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| TSLP |
| K49024_1G12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q969D9 | |
| 85480 | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title