missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSKU Monoclonal Antibody (462310)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA524182
This item is not returnable.
View return policy
Description
In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant human Tsukushi/TSK is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
The TSKU gene encodes Tsukushi, a member of the small leucine-rich proteoglycan (SLRP) family, playing crucial roles in cellular signaling, growth, and development. Tsukushi is characterized by its involvement in various biological processes, including immune regulation, tissue remodeling, and metabolic homeostasis. It is a hepatokine released in response to conditions like nonalcoholic fatty liver disease (NAFLD), influencing metabolic traits and contributing to conditions such as dyslipidemia and cardiovascular diseases. Tsukushi impacts energy expenditure by modulating brown fat thermogenesis, thereby affecting metabolic homeostasis and obesity-related metabolic traits. As an extracellular matrix component, Tsukushi participates in signaling pathways that regulate cell proliferation, migration, and differentiation, impacting organ development and pathophysiological processes. Its role in metabolic and inflammatory diseases presents it as a potential biomarker and therapeutic target in conditions characterized by altered metabolic and inflammatory states. Ongoing research further elucidates Tsukushi's mechanisms and interactions, contributing to our understanding of its multifunctional roles in health and disease.
Specifications
| TSKU | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8CBR6 | |
| TSKU | |
| Mouse myeloma cell line NS0-derived recombinant mouse Tsukushi/TSK Thr18-Leu354. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2c |
| Western Blot | |
| 462310 | |
| Unconjugated | |
| TSKU | |
| 9530051K01Rik; E2ig4; E2-induced gene 4 protein; Early insulin-induced hepatic gene protein; Eiih; hepatic protein EIIH; leucine rich repeat containing 54; leucine-rich repeat-containing protein 54; LRRC54; Tsk; Tsku; Tsukushi; tsukushi homolog; tsukushi small leucine rich proteoglycan homolog; tsukushi, small leucine rich proteoglycan; tsukushin; UNQ850/PRO1788 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 244152 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction