missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSKS Antibody (2E9), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00060385-M01
This item is not returnable.
View return policy
Description
TSKS Monoclonal antibody specifically detects TSKS in Human samples. It is validated for Western Blot, ELISA, Sandwich ELISA
Specifications
| TSKS | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| STK22 substrate 1, STK22S1, testis specific serine/threonine kinase substrate, Testis-specific kinase substrate, testis-specific serine kinase substrate, TSKS1, TSSKS | |
| TSKS (NP_068379, 471 a.a. ∽ 560 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. ALTSLVDEVKQRGLTPACPSCQRLHKKILELERQALAKHVRAEALSSTLRLAQDEALRAKNLLLTDKMKPEEKMATLDHLHLKMCSLHDH | |
| 0.1 mg | |
| Cancer, Tumor Suppressors | |
| 60385 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 2E9 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_068379 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction