missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSHZ3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599782
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TSH3.
The Teashirt zinc finger homeobox (TSHZ) family comprise a family of evolutionarily conserved transcription factors that, in Drosophila, are active in specific body parts for patterning, but whose function in vertebrates is less clear. In mice, the known three TSHZ proteins are expressed in distinct patterns in the developing and adult brain, suggesting that they play a role in the establishment of regional identity and specification of cell types within the brain. Both TSHZ3 and TSHZ1 have been found to interact with FE65, an adapter protein that binds to the amyloid protein precursor (APP) in neurons. Together with SET, a component of the inhibitor of acetyl transferase, and histone deacetylases, these proteins formed a gene-silencing complex whose target includes caspase-4. Recent experiments have also suggested this family of proteins may be involved in carcinogenesis.
Specifications
| TSHZ3 | |
| Polyclonal | |
| Unconjugated | |
| Tshz3 | |
| A630038G13Rik; Kiaa1474; mKIAA1474; teashirt 3; teashirt homolog 3; teashirt zinc finger family member 3; teashirt zinc finger homeobox 3; teashirt3; Tsh3; Tshz3; Zfp537; zinc finger protein 537; ZNF537 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 57616 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q63HK5 | |
| Tshz3 | |
| A synthesized peptide derived from human TSHZ3(Accession Q63HK5), corresponding to amino acid residues W544-P594. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction