Learn More
TSHZ2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA558255
Description
Immunogen sequence: SGSVAQLQGG NDTGTDEELE TGPEQKGCFS YQNSPGSHLS NQDAENESLL SDASDQVSDI KSVCGRDASD KKAHTHVRLP NEAHNCMDKM TAVYANILSD SYWSGLGLGF KLSNSERRNC DTRNGSNKSD FDWHQDALSK Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.
The Teashirt zinc finger homeobox (TSHZ) family comprise a family of evolutionarily conserved transcription factors that, in Drosophila, are active in specific body parts for patterning, but whose function in vertebrates is less clear. In mice, the known three TSHZ proteins are expressed in distinct patterns in the developing and adult brain, suggesting that they play a role in the establishment of regional identity and specification of cell types within the brain. Recent experiments have shown that the expression of TSHZ2 is frequently downregulated in most breast and prostate cancers and its promoter was unmethylated in virtually all cases, suggesting this family of proteins may also be involved in carcinogenesis.
Specifications
| TSHZ2 | |
| Polyclonal | |
| Unconjugated | |
| TSHZ2 | |
| 2900073F20Rik; B830045G17; C20orf17; cell growth-inhibiting protein 7; Kiaa4248; mKIAA4248; Mtsh2; Ovarian cancer-related protein 10-2; OVC10-2; Sdccag33l; SDCCAG33-like protein; serologically defined colon cancer antigen 33 like; teashirt 2; teashirt family zinc finger 2; teashirt homolog 2; teashirt homolog 2-like; teashirt zinc finger family member 2; teashirt zinc finger homeobox 2; teashirt2; TSH2; TSHZ2; ZABC2; Zfp218; Zinc finger protein 218; ZNF218 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 128553 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NRE2 | |
| TSHZ2 | |
| Recombinant protein corresponding to Human TSHZ2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Research Use Only