missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSHZ2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£285.00 - £424.00
Specifications
| Antigen | TSHZ2 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
TSHZ2 Polyclonal specifically detects TSHZ2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TSHZ2 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| C20orf17, cell growth-inhibiting protein 7, DKFZp686K2480, teashirt homolog 2, teashirt zinc finger homeobox 2, ZABC2, zinc finger protein 218 | |
| TSHZ2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 128553 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SGSVAQLQGGNDTGTDEELETGPEQKGCFSYQNSPGSHLSNQDAENESLLSDASDQVSDIKSVCGRDASDKKAHTHVRLPNEAHNCMDKMTAVYANILSDSYWSGLGLGFKLSNSERRNCDTRNGSNKSDFDWHQDALSK | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title