missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSHZ2 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82041IR
This item is not returnable.
View return policy
Description
TSHZ2 Polyclonal antibody specifically detects TSHZ2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| TSHZ2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 128553 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 755 | |
| C20orf17, cell growth-inhibiting protein 7, DKFZp686K2480, teashirt homolog 2, teashirt zinc finger homeobox 2, ZABC2, zinc finger protein 218 | |
| TSHZ2 antibody was raised against a 17 amino acid synthetic peptide near the amino terminus of human TSHZ2. The immunogen is located within amino acids 30 - 80 of TSHZ2. Amino Acid Squence: NDTGTDEELETGPEQKG | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction