missing translation for 'onlineSavingsMsg'
Learn More

TSHR Antibody (SPM223), mFluor Violet 500 SE, Novus Biologicals™

Product Code. 18421774 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421774 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421774 Supplier Novus Biologicals Supplier No. NBP254569MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TSHR Monoclonal antibody specifically detects TSHR in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSHR
Applications Flow Cytometry
Classification Monoclonal
Clone SPM223
Conjugate mFluor Violet 500 SE
Dilution Flow Cytometry
Formulation 50mM Sodium Borate
Gene Alias CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R
Host Species Mouse
Immunogen Recombinant human TSHR protein (Uniprot: P16473)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7253
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.