missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSHR Antibody (SPM222) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54317-100UG
This item is not returnable.
View return policy
Description
TSHR Monoclonal specifically detects TSHR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, CyTOF-ready.
Specifications
| TSH R | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml, CyTOF-ready | |
| CHNG1, LGR3hTSHR-I, MGC75129, seven transmembrane helix receptor, thyroid stimulating hormone receptor, Thyroid-stimulating hormone receptor, thyrotropin receptor, thyrotropin receptor-I, hTSHR-I, TSH-R | |
| Mouse | |
| 42 kDa | |
| 100 μg | |
| Cell Cycle and Replication, GPCR, Membrane Trafficking and Chaperones, Neuroscience | |
| 7253 | |
| Store at -20 to -80C. Avoid freeze-thaw cycles. | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF | |
| SPM222 | |
| Unconjugated | |
| PBS with No Preservative | |
| TSHR | |
| Recombinant human TSHR, B-Chain protein | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction