missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSH beta Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27288-1-AP-150UL
This item is not returnable.
View return policy
Description
TSH is a pituitary hormone of 28kDa, which stimulates thyroid growth and production of thyroid hormones. This antibody labels thyrotropic cells of the pituitary.Specifications
| TSH beta | |
| Polyclonal | |
| Unconjugated | |
| TSHB | |
| CHNG4, Thyrotropin alfa, Thyrotropin beta chain, Thyrotropin subunit beta, TSH B, TSH BETA, TSHB | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 7252 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P01222 | |
| TSHB | |
| TSH beta Fusion Protein Ag25532 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction