missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSH beta Rabbit anti-Human, Clone: TSHb/7001R, Novus Biologicals™
Rabbit Monoclonal Antibody
£238.00 - £444.00
Specifications
| Antigen | TSH beta |
|---|---|
| Clone | TSHb/7001R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18360534
|
Novus Biologicals
NBP3-13793-20UG |
20 μg |
£238.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18381847
|
Novus Biologicals
NBP3-13793-100UG |
100 μg |
£444.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TSH beta Monoclonal antibody specifically detects TSH beta in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| TSH beta | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA | |
| Recombinant human TSH beta fragment (Uniprot: P01222) | |
| Primary | |
| Store at 4°C. |
| TSHb/7001R | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P01222 | |
| 7252 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title