missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSH beta Monoclonal Antibody (154)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA182908
This item is not returnable.
View return policy
Description
Mouse anti Human thyroid stimulating hormone, clone 154 recognizes the beta chain of human thyroid stimulating hormone (TSH), a hormone produced by the pituitary gland, which acts to stimulate the growth of the thyroid and production of thyroid hormones.
TSH is a pituitary hormone of 28kDa, which stimulates thyroid growth and production of thyroid hormones. This antibody labels thyrotropic cells of the pituitary.
Specifications
| TSH beta | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with <0.1% sodium azide; pH 7.4 | |
| P01222 | |
| TSHB | |
| Native human beta-TSH. | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA | |
| 154 | |
| Unconjugated | |
| TSHB | |
| thyroid stimulating hormone beta; thyroid stimulating hormone subunit beta; thyroid stimulating hormone, beta; thyroid stimulating hormone, beta subunit; thyroid-stimulating hormone beta subunit; thyroid-stimulating hormone subunit beta; Thyrotropin alfa; Thyrotropin beta chain; thyrotropin beta subunit; thyrotropin subunit beta; TSH; Tshb; TSH-B; TSH-beta | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7252 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction