missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSH beta Antibody (TSH-116), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-363B
This item is not returnable.
View return policy
Description
TSH beta Monoclonal specifically detects TSH beta in Human samples. It is validated for ELISA.
Specifications
| Thyroid Stimulating Hormone | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| TSHB | |
| Human thyrotropin. | |
| 0.1 mL | |
| Cancer | |
| 7252 | |
| Human | |
| Purified |
| ELISA | |
| TSH-116 | |
| ELISA | |
| thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody (clone TSH-116) reacts with human thyroid stimulating hormone (hTSH, thyrotropin), a glycoprotein hormone produced by the anterior pituitary gland cells in response to signals from the hypothalamus gland in the brain. The TSH-116 reacts with association constant 1.1 x 1011 l/mol. Following cross-reactivity expressed as binding of labelled hormone (% of total) was determined by solid phase RIA with excess of the antibody TSH-116: hTSH (78.9), hCG (20.3), hLH (23.2), hFSH (29.9). | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction