Learn More
TSH beta Antibody (154), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-62385AF594
Product Code. 18772600
  • £578.00 / 0.50mL
Description
TSH beta Monoclonal antibody specifically detects TSH beta in Human samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TSH beta
Monoclonal
Alexa Fluor 594
50mM Sodium Borate
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry, Immunofluorescence
154
ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
thyroid stimulating hormone, beta, thyroid-stimulating hormone subunit beta, thyrotropin beta chain, thyrotropin beta subunit, thyrotropin subunit beta, TSH-B, TSH-BETA
Native Human beta-TSH
0.5 mL
Cancer, Signal Transduction
7252
Store at 4C in the dark.
IgG1