missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TSGA10IP Recombinant Mouse Monoclonal Antibody (K1E060_4H6)

Product Code. 30826299
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826299 100 μL
30827360 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826299 Supplier Invitrogen™ Supplier No. MA564438

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: CWKTEAQNLK ARQQLGAWGG VSIPTGKGEL GSEPPSGLQL PGRRPGSGSA SDKQVQLQSL GAEEAERGLS SGVLPQRPRR GSISEEEQFS EATEEAEEGE HRTPCRRRAG CQRKGQISGE EASDEGEVQG QSQGSSPSFN NLRRRQWRKT Highest antigen sequence identity to the following orthologs: Rat - 42%, Pig - 52%, Cynomolgus monkey - 86%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen TSGA10IP
Applications Western Blot
Classification Recombinant Monoclonal
Clone K1E060_4H6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene TSGA10IP
Gene Accession No. Q3SY00
Gene Alias FAM161C; testis specific 10 interacting protein; testis specific, 10 interacting protein; testis-specific protein 10-interacting protein; Tsga10-interacting protein; TSGA10IP
Gene Symbols TSGA10IP
Host Species Mouse
Immunogen Recombinant human TSGA10IP (aa 161-310).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254187, 78306
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.