missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSG101 Recombinant Rabbit Monoclonal Antibody (4T4W6)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA535689
This item is not returnable.
View return policy
Description
Immunogen sequence: MAVSESQLKK MVSKYKYRDL TVRETVNVIT LYKDLKPVLD SYVFNDGSSR ELMNLTGTIP VPYRGNTYNI PICLWLLDTY PYNPPICFVK PTSSMTIKTG.
TSG101-4A10 recognizes TSG101 protein, the product of a recently identified Tumor Susceptibility Gene whose inactivation in mouse fibroblasts results in cell transformation and the ability of those cells to form tumors in nude mice. The TSG101 gene encodes a 46 kDa protein and contains a proline-rich domain and a leucine-zipper motif within a coiled-coil domain near the carboxyl terminus.
Specifications
| TSG101 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 50% glycerol, 0.05% BSA and 0.05% ProClin 300; pH 7.3 | |
| Q61187, Q6IRE4, Q99816 | |
| Tsg101 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human TSG101/VPS23 (Q99816). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 4T4W6 | |
| Unconjugated | |
| Tsg101 | |
| AI255943; CC2; ESCRTI complex subunit TSG101; ESCRT-I complex subunit TSG101; Rw; TSG 101; TSG10; TSG101; Tumor supressor; tumor susceptibility 101; tumor susceptibility gene 10; tumor susceptibility gene 101; tumor susceptibility gene 101 protein; tumor susceptibility protein; VPS23 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 22088, 292925, 7251 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction