Learn More
TSG101 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28283-1-AP-150UL
Description
The TSG101 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TSG101. This antibody recognizes human, mouse antigen. The TSG101 antibody has been validated for the following applications: IHC, IP, WB, ELISA analysis.
Specifications
| TSG101 | |
| Polyclonal | |
| Unconjugated | |
| 150 μL | |
| 7251 | |
| IgG |
| Western Blot, Immunoprecipitation, Immunohistochemistry, ELISA | |
| 267 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |