Learn More
TSG101 Polyclonal Antibody
GREENER_CHOICE
TSG101 Polyclonal Antibody
Brand:  Thermo Scientific PA5116081
Product Code. 17238863
  • £454.00 / 100µL
Description
TSG101 Polyclonal Antibody for Western Blot, ICC/IF, IHC (PFA)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TSG101
Polyclonal
Unconjugated
Tsg101
AI255943; CC2; ESCRTI complex subunit TSG101; ESCRT-I complex subunit TSG101; Rw; TSG 101; TSG10; TSG101; Tumor supressor; tumor susceptibility 101; tumor susceptibility gene 10; tumor susceptibility gene 101; tumor susceptibility gene 101 protein; tumor susceptibility protein; VPS23
Rabbit
Affinity chromatography
RUO
22088, 292925, 7251
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
Q61187, Q6IRE4, Q99816
Tsg101
A synthesized peptide derived from human TSG101(Accession Q99816), corresponding to amino acid residues T161-T211.
100 μL
Primary
Human, Mouse, Rat, Monkey