missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSG101 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531260
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, HepG2, K562, THP-1, HL-60, Neuro 2A, C8D30, NIH-3T3, Raw264.7, C2C12, A549, H1299, HCT116, U87-MG, SK-N-SH, IMR32, SK-N-AS. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TSG101-4A10 recognizes TSG101 protein, the product of a recently identified Tumor Susceptibility Gene whose inactivation in mouse fibroblasts results in cell transformation and the ability of those cells to form tumors in nude mice. The TSG101 gene encodes a 46 kDa protein and contains a proline-rich domain and a leucine-zipper motif within a coiled-coil domain near the carboxyl terminus.
Specifications
| TSG101 | |
| Polyclonal | |
| Unconjugated | |
| Tsg101 | |
| AI255943; CC2; ESCRTI complex subunit TSG101; ESCRT-I complex subunit TSG101; Rw; TSG 101; TSG10; TSG101; Tumor supressor; tumor susceptibility 101; tumor susceptibility gene 10; tumor susceptibility gene 101; tumor susceptibility gene 101 protein; tumor susceptibility protein; VPS23 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22088, 292925, 7251 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1.42 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q61187, Q6IRE4, Q99816 | |
| Tsg101 | |
| Recombinant protein encompassing a sequence within the center region of human TSG101. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction