missing translation for 'onlineSavingsMsg'
Learn More

TSG101 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£167.00 - £388.00

Specifications

Antigen TSG101
Concentration 1.0 mg/mL
Dilution Western Blot 1 μg/ml, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/ Immunofluorescence 2 μg/ml, Immunohistochemistry-Paraffin 1:100 - 1:200
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18614819
View Documents
Novus Biologicals
NBP2-77452SS
0.025 mg
£167.00
0.03mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18657861
View Documents
Novus Biologicals
NBP2-77452
0.1 mg
£388.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TSG101 Polyclonal antibody specifically detects TSG101 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

TSG101
Western Blot 1 μg/ml, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/ Immunofluorescence 2 μg/ml, Immunohistochemistry-Paraffin 1:100 - 1:200
Polyclonal
Purified
RUO
Human, Mouse, Rat
ESCRT-I complex subunit TSG101, TSG10, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility gene 101 protein, tumor susceptibility protein, VPS23
Partial synthetic peptide made to an internal portion of the human TSG101 protein (between amino acids 125-210) [UniProt Q99816]
Immunogen affinity purified
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Autophagy, Cancer, Microautophagy
PBS
7251
Primary
Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.