missing translation for 'onlineSavingsMsg'
Learn More

TSG101 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18406955 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406955 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406955 Supplier Novus Biologicals Supplier No. NBP277452AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TSG101 Polyclonal antibody specifically detects TSG101 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSG101
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias ESCRT-I complex subunit TSG101, TSG10, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility gene 101 protein, tumor susceptibility protein, VPS23
Host Species Rabbit
Immunogen Partial synthetic peptide made to an internal portion of the human TSG101 protein (between amino acids 125-210) [UniProt Q99816]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Microautophagy
Primary or Secondary Primary
Gene ID (Entrez) 7251
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.