missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSG101 Antibody (4A10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB200-112
This item is not returnable.
View return policy
Description
TSG101 Monoclonal antibody specifically detects TSG101 in Human, Mouse, Rat, Porcine, Canine, Hamster, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunoassay, Immunohistochemistry - Free Floating, Knockdown.
Specifications
| TSG101 | |
| Monoclonal | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000, Flow Cytometry 1:25-1:200, ELISA 1:100 - 1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence 1:500-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:1000, Electron Microscopy 1:10, Immunohistochemistry Free-Floating, KnockDown Validated | |
| ESCRT-I complex subunit TSG101, TSG10, tumor susceptibility gene 10, tumor susceptibility gene 101, tumor susceptibility gene 101 protein, tumor susceptibility protein, VPS23 | |
| Mouse | |
| 44 kDa | |
| 0.1 mL | |
| Autophagy, Microautophagy | |
| 7251 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| 4A10 | |
| Unconjugated | |
| Q99816 | |
| TSG101 | |
| Amino acids 167-374 of TSG101 protein expressed in E. coli. | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Canine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction