missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ TSG General Purpose Freezers - Boxed
Shop All Thermo Scientific Products

Click to view available options
Capacity (Metric):
400 L
700 L
1500 L
Plug Type:
UK
Unit Size:
Each
Description
The Thermo Scientific™ TSG Series general purpose laboratory freezers are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.
TSG Series Freezers are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.
- Wide range of sizes from solid door 400 L to 1500 L double door.
- Whisper quite operation < 50 dBA
- White painted interior and exterior
- Adaptive Defrost
- Temperature, door ajar, and power failure (with battery backup) alarms.
- Low heat output
- Rear access port
- Keyed Lock
- LED interior lighting
- Green hydrocarbon refrigerants
- Remote alarm contacts
Specifications
Specifications
| Description | TSG Series General Purpose Laboratory Freezer |
| Capacity (Metric) | 700 L |
| Temperature Range | -12°C to -30°C |
| Temperature (Setpoint) | -20°C |
| Door Style | Solid |
| No. of Doors | 1 |
| Cabinet Material | Steel |
| Interior | Painted Steel |
| Exterior Finish | Painted Steel |
| Shelves | 4 Adjustable |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
