missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TSG General Purpose Freezers - Crated

Artikelnummer. 17137748 Alle ansehen Thermo Scientific Produkte
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
400 L
700 L
1500 L
Plug Type:
UK
missing translation for 'unitSize':
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Capacity (Metric) Plug Type missing translation for 'unitSize'
17137748 400 L UK Each
17157758 700 L UK Each
17117738 1500 L UK Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 17137748 missing translation for 'mfr' Thermo Scientific™ missing translation for 'supplierNo' TSG400FESCW

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

The TSG Series of general purpose freezers are built with the performance, security, and quality features you have come to expect from Thermo Scientific™.

The Thermo Scientific™ TSG Series general purpose laboratory freezers are designed with your needs in mind. The TSG series is ideal for the demanding requirements of daily use in research, clinical, and industrial laboratories storing reagents, media, proteins, and samples used daily in the lab.

TSG Series Freezers are designed with your laboratory in mind, combining the performance, security, and quality required for everyday laboratory applications.

  • Wide range of sizes from solid door 400 L to 1500 L double door.
  • Whisper quite operation < 50 dBA
  • White painted interior and exterior
  • Adaptive Defrost
  • Temperature, door ajar, and power failure (with battery backup) alarms.
  • Low heat output
  • Rear access port
  • Keyed Lock
  • LED interior lighting
  • Green hydrocarbon refrigerants
  • Remote alarm contacts
TRUSTED_SUSTAINABILITY

Spezifikation

Description TSG Series General Purpose Laboratory Freezer
Capacity (Metric) 400 L
Temperature Range -12°C to -30°C
Temperature (Setpoint) -20°C
Door Style Solid
No. of Doors 1
Cabinet Material Steel
Interior Painted Steel
Exterior Finish Painted Steel
Shelves 4 Adjustable
Capacity (Metric) Shelf 30 kg
Certifications/Compliance CE
Casters Yes
Control Type Digital Microprocessor
Ports 1 Rear Access Port
Warranty 12 Months
Defrost Adaptive Defrost
Packaging Type Crated
Dimensions (D x W x H) Interior 493 x 500 x 1405 mm
Dimensions (L x W x H) Exterior 493 x 500 x 1405 mm
Monitoring Options Alarm
Access Security Key Lock
Smart View Compatibility Smart-Vue Pro Compatible
Plug Type UK
Voltage 230 V
Frequency 50 Hz
Green Features Energy Efficient
Refrigerant R290
Shipping Weight (Metric) 147 kg
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.