missing translation for 'onlineSavingsMsg'
Learn More

TSFM Antibody (1A2), Novus Biologicals™

Product Code. 18379239 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379239 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379239 Supplier Novus Biologicals Supplier No. H00010102M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TSFM Monoclonal antibody specifically detects TSFM in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSFM
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1A2
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005717
Gene Alias COXPD3, EF-Ts, EFTS, EF-TsMt, EFTSMT, mitochondrial, mitochondrial elongation factor Ts, Ts translation elongation factor, mitochondrial
Host Species Mouse
Immunogen TSFM (NP_005717.2, 161 a.a. ~ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KVQWLTPVNLALWEAEAGGSLEGFLNSSELSGLPAGPDREGSLKDQLALAIGKLGENMILKRAAWVKVPSGFYVGSYVHGAMQSPSLHKLVLGKYGALVI
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Endocrinology
Primary or Secondary Primary
Gene ID (Entrez) 10102
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.