missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSEN34 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98554-50ul
This item is not returnable.
View return policy
Description
TSEN34 Polyclonal antibody specifically detects TSEN34 in Human samples. It is validated for Western Blot
Specifications
| TSEN34 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 3.1.27.9, hsSen34, leukocyte receptor cluster (LRC) member 5, Leukocyte receptor cluster member 5, PCH2C, SEN34LENG5SEN34L, tRNA splicing endonuclease 34 homolog (S. cerevisiae), tRNA splicing endonuclease 34 homolog (SEN34, S. cerevisiae), tRNA-intron endonuclease Sen34, tRNA-splicing endonuclease subunit Sen34 | |
| Produced in rabbits immunized with E. coli-derived Human TSEN34 fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 79042 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction