missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSEN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13103-2-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes one of the subunits of the tRNA splicing endonuclease. This endonuclease catalyzes the first step in RNA splicing which is the removal of introns. Mutations in this gene have been associated with pontocerebellar hypoplasia type 2. A pseudogene has been identified on chromosome 4. Multiple transcript variants encoding different isoforms have been found for this gene.Specifications
| TSEN2 | |
| Polyclonal | |
| Unconjugated | |
| TSEN2 | |
| HsSen2, PCH2B, SEN2, SEN2L, tRNA intron endonuclease Sen2, TSEN2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 312649, 381802, 80746 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.41 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5M954, Q6P7W5, Q8NCE0 | |
| Tsen2 | |
| TSEN2 Fusion Protein Ag3742 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction