missing translation for 'onlineSavingsMsg'
Learn More

TSC22/TSC22D1 Antibody (002), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18379010 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18379010 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18379010 Supplier Novus Biologicals Supplier No. NBP290141AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TSC22/TSC22D1 Monoclonal specifically detects TSC22/TSC22D1 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSC22/TSC22D1
Applications ELISA
Classification Monoclonal
Clone 2
Conjugate Alexa Fluor 405
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias Cerebral protein 2, KIAA1994, MGC17597, Regulatory protein TSC-22, RP11-269C23.2, TGFB1I4, TGFB-stimulated clone 22 homolog, transforming growth factor beta 1 induced transcript 4, Transforming growth factor beta-1-induced transcript 4 protein, transforming growth factor beta-stimulated protein TSC-22, TSC22 domain family 1, TSC22 domain family protein 1, TSC22 domain family, member 1, TSC22DKFZp686O19206
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human TSC22/TSC22D1 (Q15714-2; Met1-Ala144).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8848
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.