missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TSC2 Recombinant Rabbit Monoclonal Antibody (23GB1120), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | TSC2 |
|---|---|
| Clone | 23GB1120 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30087655
![]() |
Invitrogen™
MA554124 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088283
|
Invitrogen™
MA554125 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TSC2 Recombinant Monoclonal antibody specifically detects TSC2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western BlotSpecifications
| TSC2 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P49815, P49816, Q61037 | |
| 22084, 24855, 7249 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1120 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| LAM; Nafld; OTTHUMP00000198394; PPP1R160; protein phosphatase 1, regulatory subunit 160; Rc; renal carcinoma; Tcs2; Tsc2; TSC4; Tuberin; tuberous sclerosis 2; tuberous sclerosis 2 homolog protein; tuberous sclerosis 2 protein; Tuberous sclerosis 2 protein homolog | |
| Recombinant protein of human Tuberin (239-654AA). | |
| Primary | |
| -20°C | |
| TSC2 |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel