missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TSC2 Polyclonal antibody specifically detects TSC2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | TSC2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 50% glycerol, pH7.3 |
| Gene Alias | LAMFLJ43106, TSC4, tuberin, tuberous sclerosis 2, Tuberous sclerosis 2 protein |
| Host Species | Rabbit |
| Immunogen | A synthetic phosphorylated peptide around S1387 of human Tuberin/TSC2 (NP_000539.2). |
| Purification Method | Affinity purified |
| Quantity | 20 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?