missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC2 Rabbit anti-Human, Clone: 7X2C6, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | TSC2 |
|---|---|
| Clone | 7X2C6 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18348576
|
Novus Biologicals
NBP3-15763-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18327845
|
Novus Biologicals
NBP3-15763-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TSC2 Monoclonal antibody specifically detects TSC2 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| TSC2 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Autophagy, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Hypoxia, Inhibitors Activators and Regulators, mTOR Pathway, Signal Transduction, Tumor Suppressors | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 7249 | |
| IgG | |
| Affinity purified |
| 7X2C6 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| LAMFLJ43106, TSC4, tuberin, tuberous sclerosis 2, Tuberous sclerosis 2 protein | |
| A synthetic peptide corresponding to a sequence within amino acids 1700-1807 of human TSC2 (P49815). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title