missing translation for 'onlineSavingsMsg'
Learn More

TSC2 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18408045 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18408045 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18408045 Supplier Novus Biologicals Supplier No. NBP176430FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TSC2 Polyclonal antibody specifically detects TSC2 in Human, Mouse samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSC2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias LAMFLJ43106, TSC4, tuberin, tuberous sclerosis 2, Tuberous sclerosis 2 protein
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human TSC2. The immunogen is located within amino acids 1450 - 1500 of TSC2. Amino Acid Squence: RRGKRVERDALKSR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Hypoxia, Inhibitors Activators and Regulators, mTOR Pathway, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7249
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.