missing translation for 'onlineSavingsMsg'
Learn More

TSC1 Antibody (SR1192), R&D Systems™

Product Code. 18620815 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18620815 100 μg 100µL
18690805 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18620815 Supplier Novus Biologicals Supplier No. NBP322167100ul
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

TSC1 Monoclonal specifically detects TSC1 in Human samples. It is validated for Flow Cytometry∣Immunocytochemistry/Immunofluorescence∣Immunohistochemistry∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TSC1
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone SR1192
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Flow Cytometry 1:200, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias KIAA0243MGC86987, LAMhamartin, TSC, tuberous sclerosis 1, Tuberous sclerosis 1 protein, tumor suppressor
Host Species Rabbit
Immunogen A synthesized peptide derived from human TSC1 (Uniprot #: Q92574)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Autophagy, Cancer, Hypoxia, mTOR Pathway
Primary or Secondary Primary
Gene ID (Entrez) 7248
Target Species Human
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.