missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSC1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2315CL1
This item is not returnable.
View return policy
Description
TSC1 Polyclonal antibody specifically detects TSC1 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| TSC1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| KIAA0243MGC86987, LAMhamartin, TSC, tuberous sclerosis 1, Tuberous sclerosis 1 protein, tumor suppressor | |
| The immunogen recognized by this antibody maps to a region between residue 1100 and the C-terminus (residue 1164) of human tuberous sclerosis 1 using the numbering given in Swiss-Prot entry Q92574 (GeneID 7248). | |
| 0.1 mL | |
| Autophagy, Cancer, Hypoxia, mTOR Pathway | |
| 7248 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction