missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ TSBP Recombinant Protein Antigen
Shop All Bio Techne Products
Click to view available options
:
100 μg
Specifications
Specifications
| For Use With (Application) | Antibody Competition |
| Formulation | PBS and 1M Urea, pH 7.4. |
| Gene ID (Entrez) | 10665 |
| Molecular Weight (g/mol) | M.W. Theoretical: 27 kDa |
| Quantity | 100 μg |
| Storage Requirements | Store at -20°C. Avoid freeze-thaw cycles. |
| Gene Alias | C6orf10, chromosome 6 open reading frame 10, testis specific basic protein, TSBP |
| Gene Symbol | TSBP1 |
| Research Category | Cell Biology |
| Purity or Quality Grade | >80% by SDS-PAGE and Coomassie blue staining |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction