missing translation for 'onlineSavingsMsg'
Learn More

TSBP Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£155.00 - £364.00

Specifications

Antigen TSBP
Dilution Western Blot 1:200-1:2000
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18605032
View Documents
Novus Biologicals
NBP2-94740-0.02ml
0.02 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
18689721
View Documents
Novus Biologicals
NBP2-94740-0.1ml
0.1 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

TSBP Polyclonal antibody specifically detects TSBP in Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

TSBP
Western Blot
Unconjugated
Rabbit
Cell Biology
PBS (pH 7.3), 50% glycerol
10665
IgG
Affinity purified
Western Blot 1:200-1:2000
Polyclonal
Purified
RUO
Mouse, Rat
C6orf10, chromosome 6 open reading frame 10, testis specific basic protein, TSBP
Recombinant fusion protein containing a sequence corresponding to amino acids 220-470 of human C6orf10 (NP_006772.3). DEELAKKSCSKIQILKCGGTARSQNSREENKEALKNDIIFTNSVESLKSAHIKEPEREGKGTDLEKDKIGMEVKVDSDAGIPKRQETQLKISEMSIPQGQGAQIKKSVSDVPRGQESQVKKSESGVPKGQEAQVTKSGLVVLKGQEAQVEKSEMGVPRRQESQVKKSQSGVSKGQEAQVKKRESVVLKGQEAQVEKSELKVPKGQEGQVEKTEADVPKEQEVQEKKSEAGVLKGPESQVKNTEVSVPETLE
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documentos

Documentos

Product Certifications
Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado