missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ ClariTSA™ Fluorophore kit 650

Product Code. 18779736
Change view
Click to view available options
Quantity:
1 Kit (20 to 80 Slides)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18779736 1 Kit (20 to 80 Slides) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18779736 Supplier Tocris Bioscience™ Supplier No. 7527/1 KIT
  • ONLINE EXCLUSIVE PRICE
    £140.00
    £132.30 / 1 sets
    Save £7.70
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Signal amplification kit for use in ISH, ICC and IHC

ClariTSA™ reagents are exceptionally bright dyes that offer an effective way to boost signal intensity and detect low-abundance targets in spatial biology applications. They are designed for use in ISH, IHC and ICC experiments where in situ detection of target protein or nucleic acid sequences is required. ClariTSA reagents have been specifically optimized to deliver exceptional signal-to-noise in the RNAScope™ Multiplex Fluorescent v2 Assay for visualization of gene expression at the single cell level. They are suitable for multiplexing and can be combined with DAPI (Cat. No. 5748) counter-staining.

The basis for signal amplification comes from the specific and high-density deposition of ClariTSA dyes adjacent to a horseradish peroxidase (HRP)-labeled probe in a process termed Tyramide Signal Amplification (TSA) or Catalyzed Reporter Deposition (CARD).

Excitation maximum = 654 nm; emission maximum = 668 nm

Please see the product datasheet or protocol for information on product use.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit (20 to 80 Slides)
Target Tyramide Signaling Amplification/TSA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.