missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ TSA™ Reagent, Alexa Fluor™ 594 Tyramide
Description
New SuperBoost™ Tyramide signal Amplification Kits offer 2-10 times the sensitivity of TSA™ kits. Explore the advantages of SuperBoost™ Alexa Fluor™ 568 Tyramide Reagent, which is replacing TSA™ Reagent, Alexa Fluor™ 568 Tyramide.
This Alexa Fluor™ 594-tyramide conjugate (ex/em 591/617 nm) is a critical component of tyramide signal amplification (TSA), which is an enzyme-mediated detection method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in situ. A tyramide conjugate is incubated in the presence of HRP (often bound to a secondary antibody), which catalyzes the binding of the tyramide (and associated label) to adjacent tyrosines for enhanced fluorescent signal (if the tyramide is conjugated to a fluorescent dye) or colorimetric detection (if the tyramide is conjugated to biotin).
For ultimate detection sensitivity, tyramide-based amplification techniques are widely used in fluorescent immunodetection and FISH procedures to visualize targets with low expression levels or suboptimal fixation. It can be used with both chromogenic and fluorescent signals on slide based assays.
In addition to the stand-alone format offered here, tyramide conjugates are also available as part of an optimized kit that includes secondary antibody-HRP conjugate or streptavidin-HRP conjugate and other essential components (view a table of all available kits). For the first-time users, we recommend use of the optimized kit. However, if the primary antibody used is not compatible with the secondary antibodies available with these kits, then stand-alone tyramide conjugates (with amplification buffer) are offered that may be used as part of a customized system.
This Alexa Fluor™ 594-tyramide conjugate (ex/em 591/617 nm) is a critical component of tyramide signal amplification (TSA), which is an enzyme-mediated detection method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in situ. A tyramide conjugate is incubated in the presence of HRP (often bound to a secondary antibody), which catalyzes the binding of the tyramide (and associated label) to adjacent tyrosines for enhanced fluorescent signal (if the tyramide is conjugated to a fluorescent dye) or colorimetric detection (if the tyramide is conjugated to biotin).
For ultimate detection sensitivity, tyramide-based amplification techniques are widely used in fluorescent immunodetection and FISH procedures to visualize targets with low expression levels or suboptimal fixation. It can be used with both chromogenic and fluorescent signals on slide based assays.
In addition to the stand-alone format offered here, tyramide conjugates are also available as part of an optimized kit that includes secondary antibody-HRP conjugate or streptavidin-HRP conjugate and other essential components (view a table of all available kits). For the first-time users, we recommend use of the optimized kit. However, if the primary antibody used is not compatible with the secondary antibodies available with these kits, then stand-alone tyramide conjugates (with amplification buffer) are offered that may be used as part of a customized system.
Specifications
Specifications
| Content And Storage | -5°C to -30°C and protect from light |
| Buffer | Amplification Buffer |
| Description | TSA™ Reagent Kit, 1 Kit, Solid Form, Alexa Fluor™ 594 Label, Store at –5 to –30°C and Protect from Light, 591nm Excitation, 617nm Emission, Fluorescent Detection Method, With Amplification Buffer, Used for Cellular Imaging |
| Detection Method | Fluorescence |
| Label or Dye | Alexa Fluor 594 |
| For Use With (Application) | Cell analysis, cellular imaging, immunofluorescence (IF), immunofluorescence staining and detection, immunohistochemistry (IHC), IHC staining and detection, in situ hybridization (ISH), fluorescence in situ hybridization, immunocytochemistry (ICC) |
| Includes | Labeled tyramide, Dimethylsulfoxide (DMSO), HRP-conjugated secondary antibody or streptavidin, blocking reagent, amplification buffer, hydrogen peroxide |
| Packaging Type | 1 Kit(s) |
| Product Type | TSA™ Reagent Kit |
| Quantity | 1 Kit |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?