missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ TSA™ Reagent, Alexa Fluor™ 488 Tyramide

Product Code. 15251116
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15251116 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15251116 Supplier Molecular Probes™ Supplier No. T20948

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

TSA™ Reagent, Alexa Fluor™ 488 Tyramide

New SuperBoost™ Tyramide signal Amplification Kits offer 2-10 times the sensitivity of TSA™ kits. Explore the advantages of SuperBoost™ Alexa Fluor™ 488 Tyramide Reagent, which is replacing TSA™ Reagent, Alexa Fluor™ 488 Tyramide.

This Alexa Fluor™ 488-tyramide conjugate (ex/em 495/519 nm) is a critical component of tyramide signal amplification (TSA), which is an enzyme-mediated detection method that uses horseradish peroxidase (HRP) to generate high-density labeling of a target protein or nucleic acid in situ. A tyramide conjugate is incubated in the presence of HRP (often bound to a secondary antibody), which catalyzes the binding of the tyramide (and associated label) to adjacent tyrosines for enhanced fluorescent signal (if the tyramide is conjugated to a fluorescent dye) or colorimetric detection (if the tyramide is conjugated to biotin).

For ultimate detection sensitivity, tyramide-based amplification techniques are widely used in fluorescent immunodetection and FISH procedures to visualize targets with low expression levels or suboptimal fixation. It can be used with both chromogenic and fluorescent signals on slide based assays.

In addition to the stand-alone format offered here, tyramide conjugates are also available as part of an optimized kit that includes secondary antibody-HRP conjugate or streptavidin-HRP conjugate and other essential components (view a table of all available kits). For the first-time users, we recommend use of the optimized kit. However, if the primary antibody used is not compatible with the secondary antibodies available with these kits, then stand-alone tyramide conjugates (with amplification buffer) are offered that may be used as part of a customized system.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -5°C to -30°C
Description TSA™ Reagent, 1 kit, Solid, Alexa Fluor™ 488 Label, Used in Cellular Imaging, Protect from Light, –5 to –30°C Storage Temperature, For Research Use Only, Not For Use in Diagnostic Procedures
Label or Dye Alexa Fluor 488
For Use With (Application) Cellular Imaging
Formulation Solid
Product Type TSA™ Reagent
Quantity 1 Kit
Temperature (English) Operating 73.4 to –71.6°F
Temperature (Metric) Operating 23 to –22°C

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.