missing translation for 'onlineSavingsMsg'
Learn More

Tryptophol, 97%

Product Code. p-210157 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10434463

Brand: Thermo Scientific Acros 140600010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 526-55-6 | C10H11NO | 161.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 526-55-6
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00005659
InChI Key MBBOMCVGYCRMEA-UHFFFAOYSA-N
Synonym tryptophol, 3-2-hydroxyethyl indole, indole-3-ethanol, 1h-indole-3-ethanol, 2-1h-indol-3-yl ethanol, 3-indoleethanol, indoleethanol, 3-indolylethanol, indole ethanol, 2-3-indolyl ethanol
PubChem CID 10685
ChEBI CHEBI:17890
SMILES OCCC1=CNC2=CC=CC=C12

Specifications

Melting Point 57.0°C to 60.0°C
Color White to Brown
Boiling Point 174.0°C (2.7 mbar)
Flash Point 255°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 21, I, 218
Merck Index 15, 9978
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in glacial acetic acid, soluble in methanol, ethanol, acetone, chloroform, and ethyl acetate
IUPAC Name 2-(1H-indol-3-yl)ethanol
Formula Weight 161.2
Percent Purity 97%
Physical Form Crystalline Mass, Crystals, Flakes or Powder
Chemical Name or Material Tryptophol, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.