missing translation for 'onlineSavingsMsg'
Learn More

Tryptophol, 97%

Product Code. 10434463 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10434463 1 g Glass bottle 1g
10706171 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10434463 Supplier Thermo Scientific Acros Supplier No. 140600010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 526-55-6 | C10H11NO | 161.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 526-55-6
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.20
MDL Number MFCD00005659
InChI Key MBBOMCVGYCRMEA-UHFFFAOYSA-N
Synonym tryptophol, 3-2-hydroxyethyl indole, indole-3-ethanol, 1h-indole-3-ethanol, 2-1h-indol-3-yl ethanol, 3-indoleethanol, indoleethanol, 3-indolylethanol, indole ethanol, 2-3-indolyl ethanol
PubChem CID 10685
ChEBI CHEBI:17890
SMILES OCCC1=CNC2=CC=CC=C12

Specifications

Melting Point 57.0°C to 60.0°C
Color White to Brown
Boiling Point 174.0°C (2.7 mbar)
Flash Point 255°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Beilstein 21, I, 218
Merck Index 15, 9978
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in glacial acetic acid, soluble in methanol, ethanol, acetone, chloroform, and ethyl acetate
IUPAC Name 2-(1H-indol-3-yl)ethanol
Formula Weight 161.2
Percent Purity 97%
Physical Form Crystalline Mass, Crystals, Flakes or Powder
Chemical Name or Material Tryptophol, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.