Learn More
Tryptophanyl tRNA synthetase Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32185IR
This item is not returnable.
View return policy
Description
Tryptophanyl tRNA synthetase Polyclonal antibody specifically detects Tryptophanyl tRNA synthetase in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| Tryptophanyl tRNA synthetase | |
| Western Blot, Immunohistochemistry | |
| EC 6.1.1, EC 6.1.1.2, GAMMA-2, hWRS, IFI53, IFP53trpRS, Interferon-induced protein 53, TrpRS, tryptophan tRNA ligase 1, cytoplasmic, Tryptophan--tRNA ligase, tryptophanyl-tRNA synthetase, tryptophanyl-tRNA synthetase, cytoplasmic, WRS | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Tryptophanyl-tRNA Synthetase using the numbering given in entry NP_004175.2 (GeneID 7453). | |
| RUO |
| Western Blot, Immunohistochemistry | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Purified |