missing translation for 'onlineSavingsMsg'
Learn More

Tryptophan (L-Tryptophan), LoGiCal

Product Code. 30458924
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458924 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30458924 Supplier LoGiCal Supplier No. LGCFOR0172.00
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Tryptophan (L-Tryptophan)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-(-)-Tryptophan
CAS 73-22-3
Analyte or Component Names L-(-)-Tryptophan
Shipping Condition Room Temperature
Molecular Formula C11 H12 N2 O2
Certifications/Compliance ISO 9001
Synonym L-Tryptophan, (-)-Tryptophan, (2 S)-2-Amino-3-(1 H-indol-3-yl)propanoic acid, (S)-Tryptophan, 1 H-Indole-3-alanine, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -, 1 H-Indole-3-propanoic acid, alpha-amino-, -
InChI Formula InChI=1 S/C11H12N2O2/c12-9(11(14)15)5-7-6-13-10-4-2-1-3-8(7)10/h1-4,6,9,13 H,5,12H2,(H,14,15)/t9-/m0/s1
SMILES N[C@@H](Cc1c[nH]c2ccccc12)C(=O)O
IUPAC Name (2 S)-2-amino-3-(1 H-indol-3-yl)propanoic acid
Industry Type Forensics
Molecular Weight (g/mol) 204.23
Quantity 10 mg
Formula Weight 204.0899 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.