missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophan hydroxylase 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61104
This item is not returnable.
View return policy
Description
Tryptophan hydroxylase 2 Polyclonal specifically detects Tryptophan hydroxylase 2 in Rat samples. It is validated for Western Blot.
Specifications
| Tryptophan hydroxylase 2 | |
| Polyclonal | |
| Western Blot 1:1000 | |
| Q8CGU9 | |
| TPH2 | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser19 conjugated to KLH. Accession # Q8CGU9 | |
| Antigen Affinity-purified | |
| RUO | |
| 121278 | |
| Rat | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2 | |
| Rabbit | |
| 55 kDa | |
| 0.1 mL | |
| Primary | |
| Specific for the ∼55k tryptophan hydroxylase protein phosphorylated at Ser19. | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction