missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tryptophan hydroxylase 2 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74555IR
This item is not returnable.
View return policy
Description
Tryptophan hydroxylase 2 Polyclonal specifically detects Tryptophan hydroxylase 2 in Human, Mouse, Rat, Primate, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.
Specifications
| Tryptophan hydroxylase 2 | |
| Polyclonal | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating | |
| ADHD7, EC 1.14.16, EC 1.14.16.4, FLJ37295, MGC138872, Neuronal tryptophan hydroxylase, NTPHMGC138871, tryptophan 5-hydroxylase 2, Tryptophan 5-monooxygenase 2, tryptophan hydroxylase 2 | |
| Rabbit | |
| Affinity Purified | |
| Lipid and Metabolism, Phospho Specific | |
| 121278.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating) | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TPH2 | |
| Synthetic peptide make to an internal portion of the mouse Tryptophan hydroxylase 2 protein (between amino acids 10-75) [UniProt Q8CGV2] | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Primate, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction